Gene Transcripts

Gene Symbol
Hsd17b13
Entrez Gene ID
305150
Gene Name
hydroxysteroid (17-beta) dehydrogenase 13
Gene Aliases
Chromosome Location
Chr. 14: 7113544 - 7128218 on Build Rnor_6.0
UniGene ID
Rn.25104
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009684.1 NP_001009684.1 7 974
GenBank mRNA BC088191.1 - 7 974
FQ218851.1 - 5 571

View Full Product Details