Gene Transcripts

Gene Symbol
Itfg3
Entrez Gene ID
360502
Gene Name
integrin alpha FG-GAP repeat containing 3
Gene Aliases
RGD1311484
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.6244
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009701.1 NP_001009701.1 2526
GenBank mRNA BC088404.1 - 2526

View Full Product Details