Gene Transcripts

Gene Symbol
Mief2
Entrez Gene ID
237781
Gene Name
mitochondrial elongation factor 2
Gene Aliases
AI482195, Gm11, Smcr7
Chromosome Location
Chr. 11: 60728398 - 60732951 on Build GRCm38
UniGene ID
Mm.339760
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009927.2 NP_001009927.1 4 2260
GenBank mRNA AK156917.1 - 4 2245
AK171444.1 - 3 2384
BC088984.1 - 4 2149 5708020

View Full Product Details