Gene Transcripts

Gene Symbol
FAM83B
Entrez Gene ID
222584
Gene Name
family with sequence similarity 83 member B
Gene Aliases
C6orf143
Chromosome Location
Chr. 6: 54846323 - 54945099 on Build GRCh38
UniGene ID
Hs.152423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010872.2 NP_001010872.1 5 881
XM_006715022.3 XP_006715085.1 5 910
XM_011514394.2 XP_011512696.1 5 923
XM_011514395.2 XP_011512697.1 5 1912
XM_017010478.1 XP_016865967.1 5 907
GenBank mRNA AK055204.1 - 5 882
BC101628.1 - 4 840 8069134
BC112275.1 - 4 840 8327743

View Full Product Details