Gene Transcripts

Gene Symbol
Grwd1
Entrez Gene ID
308592
Gene Name
glutamate-rich WD repeat containing 1
Gene Aliases
Chromosome Location
Chr. 1: 101813356 - 101819058 on Build Rnor_6.0
UniGene ID
Rn.33523
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012067.1 NP_001012067.1 7 1550
GenBank mRNA BC083883.1 - 7 1550

View Full Product Details