Gene Transcripts

Gene Symbol
Dusp2
Entrez Gene ID
311406
Gene Name
dual specificity phosphatase 2
Gene Aliases
Chromosome Location
Chr. 3: 119776925 - 119779110 on Build Rnor_6.0
UniGene ID
Rn.136933
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012089.1 NP_001012089.1 4 1366
GenBank mRNA BC088205.1 - 4 1366

View Full Product Details