Gene Transcripts

Gene Symbol
Zc3h8
Entrez Gene ID
311414
Gene Name
zinc finger CCCH type containing 8
Gene Aliases
Zc3hdc8
Chromosome Location
Chr. 3: 121471836 - 121488528 on Build Rnor_6.0
UniGene ID
Rn.137486
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012090.1 NP_001012090.1 9 1383
GenBank mRNA BC079122.1 - 9 1383

View Full Product Details