Gene Transcripts

Gene Symbol
Lpin1
Entrez Gene ID
313977
Gene Name
lipin 1
Gene Aliases
Chromosome Location
Chr. 6: 41796214 - 41905152 on Build Rnor_6.0
UniGene ID
Rn.229137
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012111.1 NP_001012111.1 21 3013
XM_008764593.1 XP_008762815.1 22 2980
GenBank mRNA BC083651.1 - 21 3013

View Full Product Details