Gene Transcripts

Gene Symbol
Ptdss1
Entrez Gene ID
314553
Gene Name
phosphatidylserine synthase 1
Gene Aliases
PSS-1
Chromosome Location
Chr. 7: 71294140 - 71356153 on Build Rnor_6.0
UniGene ID
Rn.103732
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012113.1 NP_001012113.1 13 2352
GenBank mRNA BC087129.1 - 13 2352
FQ225243.1 - 6 1294

View Full Product Details