Gene Transcripts

Gene Symbol
Slc35g2
Entrez Gene ID
315957
Gene Name
solute carrier family 35, member G2
Gene Aliases
Tmem22
Chromosome Location
Chr. 8: 108854144 - 108881317 on Build Rnor_6.0
UniGene ID
Rn.51858
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012126.1 NP_001012126.1 2 1077
XM_006243630.2 XP_006243692.1 2 1245
GenBank mRNA BC088758.1 - 2 1077

View Full Product Details