Gene Transcripts

Gene Symbol
Dusp26
Entrez Gene ID
306527
Gene Name
dual specificity phosphatase 26 (putative)
Gene Aliases
RGD1310090
Chromosome Location
Chr. 16: 64798782 - 64806364 on Build Rnor_6.0
UniGene ID
Rn.22231
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012352.1 NP_001012352.1 4 1322
XM_006253232.2 XP_006253294.1 4 1170
GenBank mRNA BC089954.1 - 4 1322

View Full Product Details