Gene Transcripts

Gene Symbol
B3galnt1
Entrez Gene ID
310508
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group)
Gene Aliases
B3galt3
Chromosome Location
Chr. 2: 166226736 - 166255302 on Build Rnor_6.0
UniGene ID
Rn.3666
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013158.1 NP_001013176.1 3 821
XM_008761089.1 XP_008759311.1 1 665
GenBank mRNA BC079206.1 - 3 821

View Full Product Details