Gene Transcripts

Gene Symbol
Hn1l
Entrez Gene ID
360492
Gene Name
hematological and neurological expressed 1-like
Gene Aliases
RGD1305117
Chromosome Location
Chr. 10: 14304108 - 14324170 on Build Rnor_6.0
UniGene ID
Rn.19357
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013182.1 NP_001013200.1 5 2143
XM_006245994.2 XP_006246056.1 5 2557
GenBank mRNA BC091238.1 - 5 2143

View Full Product Details