Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013428.1 | NP_001013446.1 | 4 | 572 | |
GenBank mRNA | BC079065.1 | - | 4 | 585 | |
BC091221.1 | - | 4 | 572 | 7125035 | |
FQ227390.1 | - | 4 | 549 | ||
FQ231588.1 | - | 4 | 549 | ||
FQ234103.1 | - | 4 | 549 |