Gene Transcripts

Gene Symbol
EPGN
Entrez Gene ID
255324
Gene Name
epithelial mitogen
Gene Aliases
ALGV3072, EPG, PRO9904
Chromosome Location
Chr. 4: 74307571 - 74317298 on Build GRCh38
UniGene ID
Hs.401237
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005265666.4 XP_005265723.1 4 1521
GenBank mRNA AY358920.1 - 4 605

View Full Product Details