Gene Transcripts

Gene Symbol
Pcdh17
Entrez Gene ID
219228
Gene Name
protocadherin 17
Gene Aliases
C030033F14Rik, Gm78
Chromosome Location
Chr. 14: 84443563 - 84537565 on Build GRCm38
UniGene ID
Mm.153643
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013753.2 NP_001013775.2 4 6533
GenBank mRNA AK045478.1 - 1 524
AK046505.1 - 1 447
AK047338.1 - 1 436
AK079626.1 - 1 515
AK147265.1 - 4 6535
AK158668.1 - 4 3921
BC079850.1 - 4 4512
BC089004.1 - 4 4422 6841033

View Full Product Details