Gene Transcripts

Gene Symbol
Zscan4f
Entrez Gene ID
665902
Gene Name
zinc finger and SCAN domain containing 4F
Gene Aliases
C330039H09Rik
Chromosome Location
Chr. 7: 11397915 - 11402318 on Build GRCm38
UniGene ID
Mm.425380
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110316.2 NP_001103786.2 2 293
GenBank mRNA AK141250.1 - 2 295
CN715580.1 - 1 71

Gene Symbol
2410039M03Rik
Entrez Gene ID
70019
Gene Name
RIKEN cDNA 2410039M03 gene
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.417831
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK010645.1 - 2 345

Gene Symbol
Zscan4b
Entrez Gene ID
665780
Gene Name
zinc finger and SCAN domain containing 4B
Gene Aliases
EG665780
Chromosome Location
Chr. 7: 10900740 - 10905050 on Build GRCm38
UniGene ID
Mm.482067
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185173.1 NP_001172102.1 2 293

Gene Symbol
Zscan4c
Entrez Gene ID
245109
Gene Name
zinc finger and SCAN domain containing 4C
Gene Aliases
Gm397, XM_142517, Zscan4d
Chromosome Location
Chr. 7: 11005745 - 11010547 on Build GRCm38
UniGene ID
Mm.131747
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013765.2 NP_001013787.1 2 318
GenBank mRNA BC050218.1 - 2 318

Gene Symbol
Zscan4d
Entrez Gene ID
545913
Gene Name
zinc finger and SCAN domain containing 4D
Gene Aliases
EG545913
Chromosome Location
Chr. 7: 11160883 - 11167460 on Build GRCm38
UniGene ID
Mm.380086
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100186.1 NP_001093656.1 2 307
GenBank mRNA EF143406.1 - 2 307

View Full Product Details