Gene Transcripts

Gene Symbol
WDR6
Entrez Gene ID
11180
Gene Name
WD repeat domain 6
Gene Aliases
Chromosome Location
Chr. 3: 49007062 - 49015953 on Build GRCh38
UniGene ID
Hs.654815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320546.1 NP_001307475.1 2 862
NM_001320547.1 NP_001307476.1 2 233
NM_018031.4 NP_060501.3 2 648
GenBank mRNA AF099100.1 - 2 317
AK001080.1 - 2 317
AK225653.1 - 2 311
AK295145.1 - 2 648
AK304117.1 - 2 233
AK309913.1 - 2 528
BC002826.2 - 2 303
BC101707.1 - 2 307 8069213
BC113467.1 - 2 307 8322519
BC143486.1 - 2 307
DC300417.1 - 2 234
DC389885.1 - 2 401
DC400613.1 - 2 506
DC408104.1 - 2 429

View Full Product Details