Gene Transcripts

Gene Symbol
Clip4
Entrez Gene ID
298801
Gene Name
CAP-GLY domain containing linker protein family, member 4
Gene Aliases
RGD1311049, Rsnl2
Chromosome Location
Chr. 6: 23222020 - 23316962 on Build Rnor_6.0
UniGene ID
Rn.198194
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013942.1 NP_001013964.1 6 746
GenBank mRNA BC081910.1 - 6 746

View Full Product Details