Gene Transcripts

Gene Symbol
Cyyr1
Entrez Gene ID
304138
Gene Name
cysteine/tyrosine-rich 1
Gene Aliases
Chromosome Location
Chr. 11: 24967936 - 25078740 on Build Rnor_6.0
UniGene ID
Rn.1528
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013980.1 NP_001014002.1 4 1271
GenBank mRNA BC087052.1 - 4 1271

View Full Product Details