Gene Transcripts

Gene Symbol
Lrrc18
Entrez Gene ID
306278
Gene Name
leucine rich repeat containing 18
Gene Aliases
RGD1311738
Chromosome Location
Chr. 16: 9235512 - 9259461 on Build Rnor_6.0
UniGene ID
Rn.159999
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013999.1 NP_001014021.1 4 1120
GenBank mRNA BC081908.1 - 4 1120

View Full Product Details