Gene Transcripts
- Gene Symbol
- ARHGEF40
- Entrez Gene ID
- 55701
- Gene Name
- Rho guanine nucleotide exchange factor 40
- Gene Aliases
- SOLO
- Chromosome Location
- Chr. 14: 21061276 - 21090248 on Build GRCh38
- UniGene ID
- Hs.35125
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278529.1 |
NP_001265458.1
|
18, 19 |
4123 |
|
NM_001278530.1 |
NP_001265459.1
|
18, 19 |
3929 |
|
NM_018071.4 |
NP_060541.3
|
18, 19 |
4096 |
|
XM_005267844.2 |
XP_005267901.1
|
18, 19 |
4132 |
|
XM_011536937.2 |
XP_011535239.1
|
18, 19 |
4132 |
|
XM_017021434.1 |
XP_016876923.1
|
19, 20 |
4389 |
|
XM_017021435.1 |
XP_016876924.1
|
17, 18 |
4606 |
|
XM_017021436.1 |
XP_016876925.1
|
14, 15 |
2389 |
|
XM_017021437.1 |
XP_016876926.1
|
14, 15 |
2389 |
|
GenBank mRNA |
AK001219.1 |
- |
3, 4 |
446 |
|
AK024463.1 |
- |
16, 17 |
3334 |
|
AK074057.1 |
- |
18, 19 |
4041 |
|
AK126755.1 |
- |
13, 14 |
2683 |
|
AL137291.1 |
- |
6, 7 |
1439 |
|
BC000084.2 |
- |
4, 5 |
646 |
|
BC142692.1 |
- |
18, 19 |
4066 |
40148708 |
BC151212.1 |
- |
18, 19 |
4066 |
|
View Full Product Details