Gene Transcripts

Gene Symbol
Rab30
Entrez Gene ID
308821
Gene Name
RAB30, member RAS oncogene family
Gene Aliases
Rsb30
Chromosome Location
Chr. 1: 157573344 - 157664297 on Build Rnor_6.0
UniGene ID
Rn.161854
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015012.1 NP_001015012.1 3 860
XM_006229692.2 XP_006229754.1 5 1085
XM_006229693.2 XP_006229755.1 5 1054
XM_008759647.1 XP_008757869.1 4 4052
GenBank mRNA BC091182.1 - 3 860
FQ210359.1 - 4 961

View Full Product Details