Gene Transcripts

Gene Symbol
NDC1
Entrez Gene ID
55706
Gene Name
NDC1 transmembrane nucleoporin
Gene Aliases
NET3, TMEM48
Chromosome Location
Chr. 1: 53765460 - 53838552 on Build GRCh38
UniGene ID
Hs.476525
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168551.1 NP_001162023.1 2 442
NM_018087.4 NP_060557.3 2 442
XM_011541766.2 XP_011540068.1 2 284
GenBank mRNA AK001269.1 - 2 325
AK295000.1 - 2 226
AL354612.1 - 2 325
AL354613.1 - 2 325
BC003082.2 - 2 392 3509727
DC413103.1 - 2 442
DQ141696.1 - 2 151

View Full Product Details