Gene Transcripts

Gene Symbol
Hipk4
Entrez Gene ID
308449
Gene Name
homeodomain interacting protein kinase 4
Gene Aliases
Chromosome Location
Chr. 1: 84328114 - 84338486 on Build Rnor_6.0
UniGene ID
Rn.41008
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024776.1 NP_001019947.1 1 645
GenBank mRNA BC098070.1 - 1 645

View Full Product Details