Gene Transcripts

Gene Symbol
Prkra
Entrez Gene ID
311130
Gene Name
protein kinase, interferon inducible double stranded RNA dependent activator
Gene Aliases
RAX
Chromosome Location
Chr. 3: 63489081 - 63507918 on Build Rnor_6.0
UniGene ID
Rn.3217
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024780.1 NP_001019951.1 6, 7 732
GenBank mRNA BC097446.1 - 6, 7 732

View Full Product Details