Gene Transcripts

Gene Symbol
Slc25a39
Entrez Gene ID
360636
Gene Name
solute carrier family 25, member 39
Gene Aliases
RGD1306193
Chromosome Location
Chr. 10: 90351899 - 90356907 on Build Rnor_6.0
UniGene ID
Rn.55537
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024792.1 NP_001019963.1 12 1415
XM_006247374.2 XP_006247436.1 12 1753
XM_006247376.2 XP_006247438.1 12 1438
GenBank mRNA BC081862.1 - 12 1332
BC097349.1 - 12 1415 7460172
FQ228904.1 - 12 1407

View Full Product Details