Gene Transcripts

Gene Symbol
Mblac1
Entrez Gene ID
304346
Gene Name
metallo-beta-lactamase domain containing 1
Gene Aliases
RGD1306932
Chromosome Location
Chr. 12: 19335841 - 19337103 on Build Rnor_6.0
UniGene ID
Rn.38936
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024996.1 NP_001020167.1 1 866
GenBank mRNA BC079097.1 - 1 866

View Full Product Details