Gene Transcripts

Gene Symbol
Kdelc2
Entrez Gene ID
315664
Gene Name
KDEL (Lys-Asp-Glu-Leu) containing 2
Gene Aliases
Chromosome Location
Chr. 8: 57964988 - 57982605 on Build Rnor_6.0
UniGene ID
Rn.98686
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025123.3 NP_001020294.3 8 1746
GenBank mRNA BC093594.1 - 8 1746
BC100617.1 - 8 1745 7934552
FQ211817.1 - 7 1450

View Full Product Details