Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009978.2 | NP_034108.1 | 4 | 596 | |
GenBank mRNA | AF090691.1 | - | 4 | 585 | |
AK018943.1 | - | 4 | 597 | ||
AK054409.1 | - | 4 | 579 | ||
AK161808.1 | - | 4 | 589 | ||
BC049753.1 | - | 4 | 601 | 6773192 | |
S49926.1 | - | 4 | 397 |