Gene Transcripts

Gene Symbol
Ctag2
Entrez Gene ID
70062
Gene Name
cancer/testis antigen 2
Gene Aliases
1700030B21Rik
Chromosome Location
Chr. X: 65047644 - 65049017 on Build GRCm38
UniGene ID
Mm.45791
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027302.2 NP_081578.1 1 380
GenBank mRNA AK006526.1 - 1 381
BC049681.1 - 1 374 6703247

View Full Product Details