Gene Transcripts
- Gene Symbol
- CRTAC1
- Entrez Gene ID
- 55118
- Gene Name
- cartilage acidic protein 1
- Gene Aliases
- ASPIC, ASPIC1, CEP-68
- Chromosome Location
- Chr. 10: 97854711 - 98030828 on Build GRCh38
- UniGene ID
- Hs.500736
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001206528.2 |
NP_001193457.1
|
10, 11 |
1681 |
|
NM_018058.6 |
NP_060528.3
|
10, 11 |
1681 |
|
XM_005269938.4 |
XP_005269995.1
|
10, 11 |
1474 |
|
XM_011539917.1 |
XP_011538219.1
|
10, 11 |
1474 |
|
XM_017016366.1 |
XP_016871855.1
|
10, 11 |
1474 |
|
XM_017016367.1 |
XP_016871856.1
|
10, 11 |
1474 |
|
GenBank mRNA |
AJ276171.1 |
- |
10, 11 |
1474 |
|
AJ279016.1 |
- |
9, 10 |
1301 |
|
AJ421515.1 |
- |
9, 10 |
1301 |
|
AK001182.1 |
- |
8, 9 |
1013 |
|
AK057190.1 |
- |
10, 11 |
1681 |
|
AK292084.1 |
- |
10, 11 |
1443 |
|
BC034245.1 |
- |
8, 9 |
943 |
4178363 |
View Full Product Details