Gene Transcripts

Gene Symbol
TPD52L3
Entrez Gene ID
89882
Gene Name
tumor protein D52-like 3
Gene Aliases
NYDSP25, hD55
Chromosome Location
Chr. 9: 6328349 - 6331900 on Build GRCh38
UniGene ID
Hs.351815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001874.2 NP_001001874.1 1 381
NM_001001875.3 NP_001001875.1 1 381
NM_033516.5 NP_277051.3 1 381
XM_017015280.1 XP_016870769.1 1 369
GenBank mRNA AK057479.1 - 1 355
AY032877.1 - 1 359
BC017589.2 - 1 355 4828082
BC033792.1 - 1 326 5163651
BC042113.1 - 1 340 4838253
BI463769.1 - 398
BP369721.1 - 1 381

View Full Product Details