Gene Transcripts

Gene Symbol
Ppp4c
Entrez Gene ID
171366
Gene Name
protein phosphatase 4, catalytic subunit
Gene Aliases
Ppx
Chromosome Location
Chr. 1: 198219012 - 198225775 on Build Rnor_6.0
UniGene ID
Rn.9173
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134359.1 NP_599186.1 4, 5 492
XM_006230206.1 XP_006230268.1 5, 6 488
GenBank mRNA BC091574.1 - 4, 5 492
FQ216602.1 - 5, 6 448

View Full Product Details