Gene Transcripts

Gene Symbol
Txndc15
Entrez Gene ID
69672
Gene Name
thioredoxin domain containing 15
Gene Aliases
2310047H23Rik, AI854086, C5orf14, UNQ335
Chromosome Location
Chr. 13: 55714650 - 55726227 on Build GRCm38
UniGene ID
Mm.294457
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175150.3 NP_780359.2 4 922
GenBank mRNA AK009878.1 - 4 848
AK079517.1 - 4 869
AK160051.1 - 4 891
AK167701.1 - 4 830
AK167838.1 - 4 888
AK167977.1 - 4 887
BC029015.1 - 3 658
BC038082.1 - 2 180
BC062180.1 - 4 922 6336176
BC094275.1 - 4 758

View Full Product Details