Gene Transcripts

Gene Symbol
LMBR1L
Entrez Gene ID
55716
Gene Name
limb development membrane protein 1 like
Gene Aliases
LIMR
Chromosome Location
Chr. 12: 49096551 - 49111535 on Build GRCh38
UniGene ID
Hs.272838
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300750.1 NP_001287679.1 10, 11 1316
NM_001300751.1 NP_001287680.1 9, 10 1114
NM_018113.3 NP_060583.2 9, 10 1114
XM_011538563.2 XP_011536865.1 9, 10 1268
XM_011538564.2 XP_011536866.1 9, 10 1641
XM_011538565.2 XP_011536867.1 8, 9 1890
XM_011538566.2 XP_011536868.1 8, 9 864
XM_017019626.1 XP_016875115.1 9, 10 1516
XM_017019627.1 XP_016875116.1 10, 11 1305
XM_017019628.1 XP_016875117.1 9, 10 1103
XM_017019629.1 XP_016875118.1 10, 11 2160
XM_017019630.1 XP_016875119.1 6, 7 3076
XM_017019631.1 XP_016875120.1 8, 9 864
GenBank mRNA AB033000.1 - 9, 10 881
AF260728.1 - 9, 10 1073
AK001356.1 - 10, 11 1097
AK001651.1 - 9, 10 1064
AK093570.1 - 4, 5 900
AK298026.1 - 8, 9 1058
AK298859.1 - 3, 4 168
AK310064.1 - 10, 11 1250
AL137599.1 - 6, 7 507
AY358572.1 - 9, 10 1078
BC008337.1 - 9, 10 1044 3506347
BC015015.1 - 9, 10 843 3910520
BC031550.1 - 9, 10 875 5194023
KU178711.1 - 9, 10 769
KU178712.1 - 8, 9 752
KU178713.1 - 8, 9 735
KU178714.1 - 9, 10 769
KU178715.1 - 9, 10 769

View Full Product Details