Gene Transcripts

Gene Symbol
Hormad2
Entrez Gene ID
498400
Gene Name
HORMA domain containing 2
Gene Aliases
Chromosome Location
Chr. 14: 84576857 - 84662239 on Build Rnor_6.0
UniGene ID
Rn.138964
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017501.1 NP_001017501.1 10 1151
XM_008770344.1 XP_008768566.1 11 1312
GenBank mRNA BC079253.1 - 10 1151

View Full Product Details