Gene Transcripts

Gene Symbol
Purb
Entrez Gene ID
498407
Gene Name
purine rich element binding protein B
Gene Aliases
pur-beta
Chromosome Location
Chr. 14: 86736876 - 86739335 on Build Rnor_6.0
UniGene ID
Rn.156213
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017503.1 NP_001017503.1 1 2430
GenBank mRNA AB182574.1 - 1 2430

View Full Product Details