Gene Transcripts

Gene Symbol
Gal3st3
Entrez Gene ID
499302
Gene Name
galactose-3-O-sulfotransferase 3
Gene Aliases
Chromosome Location
Chr. 1: 220668544 - 220678300 on Build Rnor_6.0
UniGene ID
Rn.138228
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024290.1 NP_001019461.1 3 702
GenBank mRNA BC094516.1 - 3 702

View Full Product Details