Gene Transcripts
- Gene Symbol
- FRMD4A
- Entrez Gene ID
- 55691
- Gene Name
- FERM domain containing 4A
- Gene Aliases
- CCAFCA, FRMD4, bA295P9.4
- Chromosome Location
- Chr. 10: 13643706 - 14330952 on Build GRCh38
- UniGene ID
- Hs.330463
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318336.1 |
NP_001305265.1
|
7, 8 |
887 |
|
NM_001318337.1 |
NP_001305266.1
|
7, 8 |
1127 |
|
NM_018027.4 |
NP_060497.3
|
8, 9 |
827 |
|
XM_005252489.2 |
XP_005252546.1
|
7, 8 |
1127 |
|
XM_005252490.2 |
XP_005252547.1
|
7, 8 |
887 |
|
XM_006717460.2 |
XP_006717523.1
|
7, 8 |
1127 |
|
XM_011519543.2 |
XP_011517845.1
|
8, 9 |
895 |
|
XM_017016394.1 |
XP_016871883.1
|
8, 9 |
895 |
|
XM_017016395.1 |
XP_016871884.1
|
7, 8 |
730 |
|
GenBank mRNA |
AB037715.1 |
- |
8, 9 |
827 |
|
AK001072.1 |
- |
7, 8 |
895 |
|
AK094045.1 |
- |
7, 8 |
1128 |
|
AK289693.1 |
- |
8, 9 |
752 |
|
BC151244.1 |
- |
8, 9 |
838 |
8860518 |
View Full Product Details