Gene Transcripts

Gene Symbol
Fam71b
Entrez Gene ID
497888
Gene Name
family with sequence similarity 71, member B
Gene Aliases
Chromosome Location
Chr. 10: 31508549 - 31511787 on Build Rnor_6.0
UniGene ID
Rn.122734
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025031.1 NP_001020202.1 2 1908
GenBank mRNA BC082044.1 - 2 1908

View Full Product Details