Gene Transcripts

Gene Symbol
Fbxl7
Entrez Gene ID
448987
Gene Name
F-box and leucine-rich repeat protein 7
Gene Aliases
AL023057, D230018M15Rik, FBL7, Fbl6
Chromosome Location
Chr. 15: 26540454 - 26895564 on Build GRCm38
UniGene ID
Mm.253378
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176959.3 NP_795933.2 3 707
GenBank mRNA AK048688.1 - 3 721
AK129227.1 - 3 655
BC091646.1 - 3 707 30355382

View Full Product Details