Gene Transcripts

Gene Symbol
Tuba1c
Entrez Gene ID
22146
Gene Name
tubulin, alpha 1C
Gene Aliases
M[a]6, Tuba6
Chromosome Location
Chr. 15: 99029891 - 99038105 on Build GRCm38
UniGene ID
Mm.485827
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009448.4 NP_033474.1 1 319
GenBank mRNA BC094022.1 - 1 319

View Full Product Details