Gene Transcripts
- Gene Symbol
- C5orf45
- Entrez Gene ID
- 51149
- Gene Name
- chromosome 5 open reading frame 45
- Gene Aliases
- Chromosome Location
- Chr. 5: 179837266 - 179858840 on Build GRCh38
- UniGene ID
- Hs.741169
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001017987.2 |
NP_001017987.1
|
1, 2 |
118 |
|
NM_016175.3 |
NP_057259.2
|
1, 2 |
118 |
|
GenBank mRNA |
AK293901.1 |
- |
1, 2 |
84 |
|
AK295104.1 |
- |
1, 2 |
111 |
|
AK298954.1 |
- |
1, 2 |
75 |
|
BC056889.1 |
- |
1, 2 |
71 |
|
BC069051.1 |
- |
1, 2 |
86 |
3945859 |
BF965313.1 |
- |
1, 2 |
119 |
|
BQ642639.1 |
- |
1, 2 |
74 |
|
BX537968.1 |
- |
1, 2 |
139 |
|
BX647375.1 |
- |
1, 2 |
86 |
|
CN363239.1 |
- |
1, 2 |
42 |
|
CR936785.1 |
- |
1, 2 |
139 |
|
View Full Product Details