Gene Transcripts

Gene Symbol
Ly6g6d
Entrez Gene ID
114654
Gene Name
lymphocyte antigen 6 complex, locus G6D
Gene Aliases
A930024F17Rik, G6d, G6f, MEGT1, NG25, NG32
Chromosome Location
Chr. 17: 35071342 - 35080567 on Build GRCm38
UniGene ID
Mm.246697
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033478.2 NP_258439.1 3 487
GenBank mRNA BB737333.1 - 1 283

View Full Product Details