Gene Transcripts

Gene Symbol
PLXNB2
Entrez Gene ID
23654
Gene Name
plexin B2
Gene Aliases
MM1, Nbla00445, PLEXB2, dJ402G11.3
Chromosome Location
Chr. 22: 50274979 - 50307637 on Build GRCh38
UniGene ID
Hs.3989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012401.3 NP_036533.2 36 6160
XM_005261909.1 XP_005261966.1 37 6208
XM_005261910.2 XP_005261967.1 37 6219
XM_005261911.4 XP_005261968.1 36 6158
XM_006724413.2 XP_006724476.1 38 6278
XM_011530682.2 XP_011528984.1 37 6359
XM_017028703.1 XP_016884192.1 38 6259
XM_017028704.1 XP_016884193.1 37 6212
GenBank mRNA AB002313.1 - 35 6077
AB075511.1 - 21 3270
AK025415.1 - 11 1747
AK025701.1 - 23 3528
AK056543.1 - 19 3329
AK074932.1 - 18 2796
AK123131.1 - 8 1741
AK126394.1 - 17 2869
BC004542.2 - 13 1979
BC110366.1 - 8 1397

View Full Product Details