Gene Transcripts

Gene Symbol
COA1
Entrez Gene ID
55744
Gene Name
cytochrome c oxidase assembly factor 1 homolog
Gene Aliases
C7orf44, MITRAC15
Chromosome Location
Chr. 7: 43608452 - 43729541 on Build GRCh38
UniGene ID
Hs.268887
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321197.1 NP_001308126.1 5 507
NM_001321198.1 NP_001308127.1 5 388
NM_001321199.1 NP_001308128.1 5 352
NM_001321200.1 NP_001308129.1 5 408
NM_001321201.1 NP_001308130.1 4 280
NM_018224.3 NP_060694.2 4 287
NR_047701.1 5 352
NR_135580.1 5 388
NR_135581.1 5 388
NR_135582.1 4 287
NR_135583.1 4 287
XM_005249807.4 XP_005249864.1 4 351
XM_006715752.2 XP_006715815.1 5 319
XM_017012413.1 XP_016867902.1 4 278
XM_017012414.1 XP_016867903.1 4 278
XM_017012415.1 XP_016867904.1 5 380
XM_017012416.1 XP_016867905.1 4 278
XM_017012417.1 XP_016867906.1 4 278
XM_017012418.1 XP_016867907.1 4 278
XM_017012419.1 XP_016867908.1 4 278
XM_017012420.1 XP_016867909.1 5 380
XM_017012421.1 XP_016867910.1 5 502
XM_017012422.1 XP_016867911.1 5 391
XM_017012423.1 XP_016867912.1 5 311
XM_017012424.1 XP_016867913.1 4 294
XM_017012425.1 XP_016867914.1 5 395
XM_017012426.1 XP_016867915.1 5 514
XM_017012427.1 XP_016867916.1 6 469
XM_017012428.1 XP_016867917.1 4 294
XM_017012429.1 XP_016867918.1 3 188
XM_017012430.1 XP_016867919.1 5 380
XM_017012431.1 XP_016867920.1 4 294
XM_017012432.1 XP_016867921.1 4 294
GenBank mRNA AK001665.1 - 5 321
AK021905.1 - 5 457
AK023007.1 - 5 304
AK023053.1 - 5 320
AK097203.1 - 3 1818
AK293043.1 - 5 330
BC056884.1 - 5 279 6159045
BG028456.1 - 309
BG548154.1 - 5 344
BM918741.1 - 4 260
BP373150.1 - 6 525
BP377872.1 - 4 288
BQ231567.1 - 4 225
DA742258.1 - 4 269

View Full Product Details