Gene Transcripts
- Gene Symbol
- ATAD3C
- Entrez Gene ID
- 219293
- Gene Name
- ATPase family, AAA domain containing 3C
- Gene Aliases
- Chromosome Location
- Chr. 1: 1449689 - 1470158 on Build GRCh38
- UniGene ID
- Hs.724767
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK074653.1 |
- |
1 |
944 |
|
View Full Product Details