Gene Transcripts
- Gene Symbol
- ASUN
- Entrez Gene ID
- 55726
- Gene Name
- asunder, spermatogenesis regulator
- Gene Aliases
- C12orf11, GCT1, Mat89Bb, NET48, SPATA30
- Chromosome Location
- Chr. 12: 26905171 - 26938326 on Build GRCh38
- UniGene ID
- Hs.505077
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018164.2 |
NP_060634.2
|
3, 4 |
842 |
|
XM_005253422.3 |
XP_005253479.1
|
3, 4 |
801 |
|
XM_011520750.2 |
XP_011519052.1
|
3, 4 |
375 |
|
XM_011520751.2 |
XP_011519053.1
|
3, 4 |
801 |
|
XM_011520752.2 |
XP_011519054.1
|
3, 4 |
801 |
|
XM_017019632.1 |
XP_016875121.1
|
3, 4 |
375 |
|
XM_017019635.1 |
XP_016875124.1
|
3, 4 |
375 |
|
XM_017019636.1 |
XP_016875125.1
|
3, 4 |
801 |
|
GenBank mRNA |
AK001492.1 |
- |
3, 4 |
471 |
|
AK001499.1 |
- |
3, 4 |
507 |
|
AK225336.1 |
- |
3, 4 |
493 |
|
BC003081.2 |
- |
3, 4 |
531 |
2989678 |
BC008368.1 |
- |
3, 4 |
489 |
3355282 |
View Full Product Details