Gene Transcripts

Gene Symbol
FAM87A
Entrez Gene ID
157693
Gene Name
family with sequence similarity 87 member A
Gene Aliases
Chromosome Location
Chr. 8: 375931 - 383174 on Build GRCh38
UniGene ID
Hs.591390
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_103537.1 4 3902
GenBank mRNA BC037297.2 - 4 3901

Gene Symbol
FAM87B
Entrez Gene ID
400728
Gene Name
family with sequence similarity 87 member B
Gene Aliases
Chromosome Location
Chr. 1: 817371 - 819834 on Build GRCh38
UniGene ID
Hs.568078
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_103536.1 2 1836
GenBank mRNA AK097327.1 - 2 1836

View Full Product Details